Assay Details
| Gene Symbol: AT4G17580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117865.1 | NP_193492.1 | 4 - 5 | 360 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G17580| Gene Symbol: AT4G17580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117865.1 | NP_193492.1 | 4 - 5 | 360 | 71 | |
Set Membership: |
|