Assay Details
| Gene Symbol: CKB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203828.2 | NP_001190757.1 | 4 - 5 | 992 | 80 | |
| NM_117872.3 | NP_193499.1 | 4 - 5 | 1007 | 80 | ||
| GenBank mRNA | AK119067.1 | - | 4 - 5 | 794 | 80 | |
| AY085899.1 | - | 4 - 5 | 955 | 80 | ||
| BT005279.1 | - | 4 - 5 | 714 | 80 | ||
| U03984.1 | - | 4 - 5 | 847 | 80 | ||
More Information
Set Membership: |
|

CKB2
Genomic Map