Assay Details
| Gene Symbol: SYP23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117882.3 | NP_567537.1 | 6 - 7 | 1022 | 112 | |
| GenBank mRNA | AY074294.1 | - | 6 - 7 | 980 | 112 | |
| BT000708.1 | - | 6 - 7 | 979 | 112 | ||
| U85036.1 | - | 6 - 7 | 949 | 112 | ||
More Information
Set Membership: |
|

SYP23
Genomic Map