Assay Details
| Gene Symbol: AT4G17940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117904.4 | NP_567545.1 | 2 - 3 | 753 | 111 | |
| GenBank mRNA | AK226990.1 | - | 2 - 3 | 751 | 111 | |
More Information
Set Membership: |
|

AT4G17940
Genomic Map