Assay Details
| Gene Symbol: SSP4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036584.3 | NP_001031661.2 | 2 - 3 | 304 | 108 | |
| NM_001203831.1 | NP_001190760.1 | 2 - 3 | 304 | 108 | ||
| NM_001341261.1 | NP_001328767.1 | 2 - 3 | 345 | 108 | ||
| NM_001341262.1 | NP_001328766.1 | 2 - 3 | 304 | 108 | ||
| NM_001341263.1 | NP_001328768.1 | 1 - 2 | 571 | 108 | ||
| NM_117924.7 | NP_193548.7 | 1 - 2 | 527 | 108 | ||
More Information
Set Membership: |
|

SSP4b
Genomic Map