Assay Details
| Gene Symbol: SS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117934.4 | NP_193558.3 | 13 - 14 | 2923 | 93 | |
| GenBank mRNA | AK226610.1 | - | 13 - 14 | 2928 | 93 | |
| KJ676990.1 | - | 13 - 14 | 2810 | 93 | ||
More Information
Set Membership: |
|

SS4
Genomic Map