Assay Details
| Gene Symbol: SNI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117959.1 | NP_567557.1 | 12 - 13 | 1044 | 141 | |
| GenBank mRNA | AF169596.1 | - | 12 - 13 | 1028 | 141 | |
| BT029212.1 | - | 12 - 13 | 969 | 141 | ||
More Information
Set Membership: |
|

SNI1
Genomic Map