Assay Details
| Gene Symbol: AT4G18465 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117961.3 | NP_567558.2 | 12 - 13 | 1150 | 150 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G18465| Gene Symbol: AT4G18465 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117961.3 | NP_567558.2 | 12 - 13 | 1150 | 150 | |
Set Membership: |
|