Assay Details
| Gene Symbol: RUG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343030.1 | NP_001332124.1 | 11 - 12 | 1513 | 63 | |
| NM_147851.4 | NP_680156.2 | 12 - 13 | 1445 | 63 | ||
| GenBank mRNA | AK118882.1 | - | 12 - 13 | 1332 | 63 | |
| BT005498.1 | - | 12 - 13 | 1097 | 63 | ||
More Information
Set Membership: |
|

RUG1
Genomic Map