Assay Details
| Gene Symbol: AT5G08720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147852.4 | NP_680157.1 | 8 - 9 | 2025 | 83 | |
| GenBank mRNA | AK117869.1 | - | 8 - 9 | 1876 | 83 | |
| BT005959.1 | - | 8 - 9 | 1830 | 83 | ||
More Information
Set Membership: |
|

AT5G08720
Genomic Map