Assay Details
| Gene Symbol: PAA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036843.1 | NP_001031920.1 | 12 - 13 | 1731 | 69 | |
| NM_001203428.1 | NP_001190357.1 | 11 - 12 | 1698 | 69 | ||
| NM_001343706.1 | NP_001331520.1 | 12 - 13 | 1754 | 69 | ||
| NM_147876.5 | NP_680181.2 | 12 - 13 | 1754 | 69 | ||
| GenBank mRNA | AK316941.1 | - | 12 - 13 | 1665 | 69 | |
More Information
Set Membership: |
|

PAA2
Genomic Map