Assay Details
| Gene Symbol: PAA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036843.1 | NP_001031920.1 | 13 - 14 | 2368 | 155 | |
| NM_001203428.1 | NP_001190357.1 | 12 - 13 | 2335 | 155 | ||
| NM_001343706.1 | NP_001331520.1 | 13 - 14 | 2391 | 155 | ||
| NM_147876.5 | NP_680181.2 | 13 - 14 | 2391 | 155 | ||
| GenBank mRNA | AK316941.1 | - | 13 - 14 | 2302 | 155 | |
More Information
Set Membership: |
|

PAA2
Genomic Map