Assay Details
| Gene Symbol: UBP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343718.1 | NP_001318617.1 | 1 - 2 | 93 | 62 | |
| NM_122126.5 | NP_568411.4 | 1 - 2 | 93 | 62 | ||
More Information
Set Membership: |
|

UBP8
Genomic Map