Assay Details
| Gene Symbol: PNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343720.1 | NP_001332623.1 | 1 - 2 | 284 | 117 | |
| NM_147894.4 | NP_680199.1 | 2 - 3 | 523 | 117 | ||
| GenBank mRNA | AK226614.1 | - | 2 - 3 | 528 | 117 | |
| AY302133.1 | - | 1 - 2 | 276 | 117 | ||
| BT022100.1 | - | 2 - 3 | 428 | 117 | ||
| BT026521.1 | - | 2 - 3 | 302 | 117 | ||
| KJ138673.1 | - | 2 - 3 | 302 | 117 | ||
More Information
Set Membership: |
|

PNT1
Genomic Map