Assay Details
| Gene Symbol: EMB1030 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343771.1 | NP_001330952.1 | 5 - 6 | 2313 | 74 | |
| NM_147905.3 | NP_680210.2 | 5 - 6 | 2351 | 74 | ||
More Information
Set Membership: |
|

EMB1030
Genomic Map