Assay Details
| Gene Symbol: ECI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105226.4 | NP_176730.1 | 1 | 854 | 144 | |
| GenBank mRNA | BT010695.1 | - | 1 | 793 | 144 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ECI1| Gene Symbol: ECI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105226.4 | NP_176730.1 | 1 | 854 | 144 | |
| GenBank mRNA | BT010695.1 | - | 1 | 793 | 144 | |
Set Membership: |
|