Assay Details
| Gene Symbol: FRA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334216.1 | NP_001321558.1 | 10 - 11 | 3106 | 77 | |
| NM_105232.3 | NP_176736.2 | 10 - 11 | 3089 | 77 | ||
| GenBank mRNA | AY761186.1 | - | 10 - 11 | 3062 | 77 | |
More Information
Set Membership: |
|

FRA3
Genomic Map