Assay Details
| Gene Symbol: UCH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105238.3 | NP_564858.1 | 5 - 6 | 424 | 84 | |
| GenBank mRNA | AY065083.1 | - | 5 - 6 | 347 | 84 | |
| AY114549.1 | - | 4 - 5 | 225 | 84 | ||
More Information
Set Membership: |
|

UCH2
Genomic Map