Assay Details
| Gene Symbol: AT1G65990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105271.1 | NP_176774.1 | 1 - 2 | 154 | 70 | |
| GenBank mRNA | DQ056510.1 | - | 1 - 2 | 154 | 70 | |
More Information
Set Membership: |
|

AT1G65990
Genomic Map