Assay Details
| Gene Symbol: CMPG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105287.2 | NP_564866.1 | 1 | 235 | 78 | |
| GenBank mRNA | AY034913.1 | - | 1 | 232 | 78 | |
| AY063104.1 | - | 1 | 213 | 78 | ||
More Information
Set Membership: |
|

CMPG1
Genomic Map