Assay Details
| Gene Symbol: MMD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105288.2 | NP_176791.2 | 2 - 3 | 811 | 78 | |
| GenBank mRNA | AY158082.1 | - | 2 - 3 | 811 | 78 | |
| AY305007.1 | - | 2 - 3 | 610 | 78 | ||
More Information
Set Membership: |
|

MMD1
Genomic Map