Assay Details
| Gene Symbol: GSR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036163.2 | NP_001031240.1 | 1 - 2 | 191 | 84 | |
| NM_001198398.1 | NP_001185327.1 | 1 - 2 | 236 | 84 | ||
| NM_105291.4 | NP_176794.1 | 1 - 2 | 256 | 84 | ||
| GenBank mRNA | AY091101.1 | - | 1 - 2 | 164 | 84 | |
| AY122962.1 | - | 1 - 2 | 73 | 84 | ||
More Information
Set Membership: |
|

GSR2
Genomic Map