Assay Details
| Gene Symbol: AT1G66430 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105314.5 | NP_564875.2 | 3 - 4 | 706 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G66430| Gene Symbol: AT1G66430 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105314.5 | NP_564875.2 | 3 - 4 | 706 | 91 | |
Set Membership: |
|