Assay Details
| Gene Symbol: RHD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105318.3 | NP_176820.1 | 4 - 5 | 882 | 92 | |
| GenBank mRNA | AY128310.1 | - | 4 - 5 | 883 | 92 | |
| BT006320.1 | - | 4 - 5 | 770 | 92 | ||
More Information
Set Membership: |
|

RHD6
Genomic Map