Assay Details
| Gene Symbol: CLPP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105338.4 | NP_564880.1 | 2 - 3 | 619 | 141 | |
| GenBank mRNA | AB022328.1 | - | 2 - 3 | 418 | 141 | |
| AY087349.1 | - | 2 - 3 | 471 | 141 | ||
More Information
Set Membership: |
|

CLPP3
Genomic Map