Assay Details
| Gene Symbol: CLPP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105338.4 | NP_564880.1 | 4 - 5 | 889 | 144 | |
| GenBank mRNA | AB022328.1 | - | 4 - 5 | 688 | 144 | |
| AY087349.1 | - | 4 - 5 | 741 | 144 | ||
More Information
Set Membership: |
|

CLPP3
Genomic Map