Assay Details
| Gene Symbol: AR401 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105339.4 | NP_176841.1 | 6 - 7 | 1025 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AR401| Gene Symbol: AR401 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105339.4 | NP_176841.1 | 6 - 7 | 1025 | 69 | |
Set Membership: |
|