Assay Details
| Gene Symbol: CAK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105345.5 | NP_176847.1 | 2 - 3 | 652 | 110 | |
| GenBank mRNA | AB074116.1 | - | 2 - 3 | 439 | 110 | |
| AY136355.1 | - | 2 - 3 | 426 | 110 | ||
| BT000198.1 | - | 2 - 3 | 400 | 110 | ||
More Information
Set Membership: |
|

CAK4
Genomic Map