Assay Details
| Gene Symbol: AT1G67120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334273.1 | NP_001322748.1 | 12 - 13 | 1739 | 91 | |
| NM_105382.7 | NP_176883.5 | 12 - 13 | 1851 | 91 | ||
More Information
Set Membership: |
|

AT1G67120
Genomic Map