Assay Details
| Gene Symbol: TEL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105445.2 | NP_176943.1 | 2 - 3 | 1224 | 113 | |
| GenBank mRNA | DQ446408.1 | - | 2 - 3 | 1045 | 113 | |
More Information
Set Membership: |
|

TEL2
Genomic Map