Assay Details
| Gene Symbol: ANN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334345.1 | NP_001320670.1 | 2 - 3 | 340 | 67 | |
| NM_105479.2 | NP_564920.1 | 2 - 3 | 340 | 67 | ||
| GenBank mRNA | DQ446409.1 | - | 2 - 3 | 219 | 67 | |
More Information
Set Membership: |
|

ANN5
Genomic Map