Assay Details
| Gene Symbol: AT1G68570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105528.5 | NP_177024.1 | 1 - 2 | 221 | 76 | |
| GenBank mRNA | AK226963.1 | - | 1 - 2 | 183 | 76 | |
| AY091784.1 | - | 1 - 2 | 181 | 76 | ||
| BT002278.1 | - | 1 - 2 | 107 | 76 | ||
More Information
Set Membership: |
|

AT1G68570
Genomic Map