Assay Details
| Gene Symbol: AT1G68570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334368.1 | NP_001321309.1 | 1 - 2 | 233 | 73 | |
| NM_105528.5 | NP_177024.1 | 2 - 3 | 437 | 73 | ||
| GenBank mRNA | AK226963.1 | - | 2 - 3 | 399 | 73 | |
| AY091784.1 | - | 2 - 3 | 397 | 73 | ||
| BT002278.1 | - | 2 - 3 | 323 | 73 | ||
More Information
Set Membership: |
|

AT1G68570
Genomic Map