Assay Details
| Gene Symbol: AT1G68570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334368.1 | NP_001321309.1 | 2 - 3 | 468 | 87 | |
| NM_105528.5 | NP_177024.1 | 3 - 4 | 672 | 87 | ||
| GenBank mRNA | AK226963.1 | - | 3 - 4 | 634 | 87 | |
| AY091784.1 | - | 3 - 4 | 632 | 87 | ||
| BT002278.1 | - | 3 - 4 | 558 | 87 | ||
More Information
Set Membership: |
|

AT1G68570
Genomic Map