Assay Details
| Gene Symbol: AT1G68630 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105535.4 | NP_177030.4 | 3 - 4 | 521 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G68630| Gene Symbol: AT1G68630 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105535.4 | NP_177030.4 | 3 - 4 | 521 | 80 | |
Set Membership: |
|