Assay Details
| Gene Symbol: PAN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105536.4 | NP_177031.1 | 1 - 2 | 418 | 146 | |
| GenBank mRNA | AK118286.1 | - | 1 - 2 | 239 | 146 | |
| BT008586.1 | - | 1 - 2 | 71 | 146 | ||
More Information
Set Membership: |
|

PAN
Genomic Map