Assay Details
| Gene Symbol: PAN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105536.4 | NP_177031.1 | 4 - 5 | 767 | 121 | |
| GenBank mRNA | AK118286.1 | - | 4 - 5 | 588 | 121 | |
| BT008586.1 | - | 4 - 5 | 420 | 121 | ||
More Information
Set Membership: |
|

PAN
Genomic Map