Assay Details
| Gene Symbol: AT1G68730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105545.3 | NP_177040.2 | 1 - 2 | 211 | 81 | |
| GenBank mRNA | AB493527.1 | - | 1 - 2 | 159 | 81 | |
More Information
Set Membership: |
|

AT1G68730
Genomic Map