Assay Details
| Gene Symbol: AT1G68730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105545.3 | NP_177040.2 | 2 - 3 | 333 | 137 | |
| GenBank mRNA | AB493527.1 | - | 2 - 3 | 281 | 137 | |
More Information
Set Membership: |
|

AT1G68730
Genomic Map