Assay Details
| Gene Symbol: SOFL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105560.3 | NP_177053.1 | 1 | 546 | 122 | |
| GenBank mRNA | BT014768.1 | - | 1 | 520 | 122 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SOFL2| Gene Symbol: SOFL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105560.3 | NP_177053.1 | 1 | 546 | 122 | |
| GenBank mRNA | BT014768.1 | - | 1 | 520 | 122 | |
Set Membership: |
|