Assay Details
| Gene Symbol: WIT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084328.2 | NP_001077797.1 | 2 - 3 | 599 | 87 | |
| NM_001198424.2 | NP_001185353.1 | 2 - 3 | 722 | 87 | ||
| NM_105565.3 | NP_177057.2 | 2 - 3 | 553 | 87 | ||
More Information
Set Membership: |
|

WIT2
Genomic Map