Assay Details
| Gene Symbol: WIT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084328.2 | NP_001077797.1 | 3 - 4 | 1706 | 123 | |
| NM_001198424.2 | NP_001185353.1 | 3 - 4 | 1829 | 123 | ||
| NM_105565.3 | NP_177057.2 | 3 - 4 | 1660 | 123 | ||
More Information
Set Membership: |
|

WIT2
Genomic Map