Assay Details
| Gene Symbol: AT1G68920 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036178.1 | NP_001031255.1 | 3 - 4 | 1637 | 88 | |
| NM_001334390.1 | NP_001320380.1 | 4 - 5 | 1555 | 88 | ||
| NM_105566.5 | NP_177058.1 | 3 - 4 | 1663 | 88 | ||
| NM_179532.4 | NP_849863.2 | 4 - 5 | 1437 | 88 | ||
| GenBank mRNA | AK318937.1 | - | 4 - 5 | 1359 | 88 | |
| AY128299.1 | - | 3 - 4 | 1635 | 88 | ||
| BT002259.1 | - | 3 - 4 | 1027 | 88 | ||
More Information
Set Membership: |
|

AT1G68920
Genomic Map