Assay Details
| Gene Symbol: MGP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198426.1 | NP_001185355.1 | 11 - 12 | 2343 | 73 | |
| NM_105571.4 | NP_177063.1 | 11 - 12 | 2393 | 73 | ||
More Information
Set Membership: |
|

MGP3
Genomic Map