Assay Details
| Gene Symbol: MGP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198426.1 | NP_001185355.1 | 13 - 14 | 2574 | 70 | |
| NM_105571.4 | NP_177063.1 | 13 - 14 | 2624 | 70 | ||
More Information
Set Membership: |
|

MGP3
Genomic Map