Assay Details
| Gene Symbol: MGP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198426.1 | NP_001185355.1 | 2 - 3 | 1153 | 135 | |
| NM_105571.4 | NP_177063.1 | 2 - 3 | 1224 | 135 | ||
More Information
Set Membership: |
|

MGP3
Genomic Map