Assay Details
| Gene Symbol: MGP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198426.1 | NP_001185355.1 | 3 - 4 | 1294 | 100 | |
| NM_105571.4 | NP_177063.1 | 3 - 4 | 1365 | 100 | ||
More Information
Set Membership: |
|

MGP3
Genomic Map