Assay Details
| Gene Symbol: MGP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105571.4 | NP_177063.1 | 4 - 5 | 1431 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGP3| Gene Symbol: MGP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105571.4 | NP_177063.1 | 4 - 5 | 1431 | 85 | |
Set Membership: |
|