Assay Details
| Gene Symbol: MGP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198426.1 | NP_001185355.1 | 8 - 9 | 1805 | 161 | |
| NM_105571.4 | NP_177063.1 | 8 - 9 | 1855 | 161 | ||
More Information
Set Membership: |
|

MGP3
Genomic Map