Assay Details
| Gene Symbol: BIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105572.5 | NP_177064.1 | 4 - 5 | 536 | 107 | |
| GenBank mRNA | BT002352.1 | - | 4 - 5 | 361 | 107 | |
More Information
Set Membership: |
|

BIM2
Genomic Map